Learn More
Mettler Toledo™ LE621 Sensor for FiveGo™ Dissolved Oxygen Meters
Catalog No. 01912369 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Length (Metric) Cable:
1 m
Catalog No. Length (Metric) Cable
01-912-369 1 m
1 options
Catalog No. 01-912-369 Supplier Mettler Toledo™ Supplier No. 30266975
Add to Cart
Add to Cart
IP67 waterproof, galvanic dissolved oxygen sensor with integrated temperature sensor.
  • Robust and easy to use
  • Plug/Connection: BNC/Cinch
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC/RCA (Cinch) IP67
Description LE DO Electrode LE621-IP67
Type LE621 Sensor, IP67 Waterproof
Includes Integrated temperature sensor
Length (Metric) Cable 1 m