Learn More
Mettler Toledo™ LE pH Electrode LE438-IP67 pH/Conductivity/DO Meter
Catalog No. 01912366 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
FiveGO, FiveEasy, and other Mettler Toledo pH Meters
Catalog No. For Use With (Equipment)
01-912-366 FiveGO, FiveEasy, and other Mettler Toledo pH Meters
1 options
Catalog No. 01-912-366 Supplier Mettler Toledo™ Supplier No. 30247153
Add to Cart
Add to Cart
IP67 waterproof, plastic pH electrode with integrated temperature sensor.
  • Low maintainence
  • For field use and for samples with fluctuating temperatures
  • Plug/Connection: BNC/Cinch
Order Info
Measures pH only.
TRUSTED_SUSTAINABILITY
Specifications
Type LE438 Sensor; IP67 Waterproof
For Use With (Application) Field use, samples with fluctuating temperatures
Includes Integrated temperature sensor
For Use With (Equipment) FiveGO, FiveEasy, and other Mettler Toledo pH Meters