Learn More
Velp Scientifica™ FIWE3 Raw Fiber Extractor
SDP
Catalog No. 07222019
Change view
Click to view available options
For Use With (Equipment):
Glass Crucibles
Catalog No. For Use With (Equipment)
07-222-019 Glass Crucibles
1 options
This item is not returnable. View return policy
Catalog No. 07-222-019 Supplier Velp Scientifica™ Supplier No. SA30540201
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
This fiber analyzer has 3 positions that use pre-heated reagents for fiber analysis according to Weende, Van Soest and other official methods.
  • Suitable for raw fiber extraction, conventionally known as an indigestible residue
  • Rapid analysis, reliable results and high reproducibility are some of the most relevant benefits of these units which are ideal for the following applications: the Weende method, and the Van Soest method
  • Perform single or multiple extractions including boiling, rinsing and filtration steps with the maximum reproducibility available
  • No sample transfer prevents any possible sample loss, as crucibles can also be used as sample vessels during weighing, drying and washing
  • Reduces the time required for the analysis: the equivalent manual procedure will require more than 6 hours, compared to the 2 hours with FIWE 3
Specifications
Product Type Fiber Extractor
For Use With (Equipment) Glass Crucibles
Capacity (Metric) 3 g
Voltage 115 V