Learn More
Electron Microscopy Sciences Fix-'N'-Temp Container
SDP
Catalog No. 5025948
Encompass_Preferred
This container will permit rapid specimen handling and provides temperature information during microwave irradiation. A liquid crystal strip is affixed into the bottom of a 35 mm diameter petri dish and covered with a thin layer of EMbed resin. The temperature range is 35 degree C-60 degree C. This container is ideal for tissue fixing by microwaves. The two active temperature windows are 45 degree C and 50 degree C.
Catalog No. 50-259-48 Supplier Electron Microscopy Sciences Supplier No. 9703301
This container will permit rapid specimen handling and provides temperature information during microwave irradiation. A liquid crystal strip is affixed into the bottom of a 35 mm diameter petri dish and covered with a thin layer of EMbed resin. The temperature range is 35 degree C-60 degree C. This container is ideal for tissue fixing by microwaves. The two active temperature windows are 45 degree C and 50 degree C.