Learn More
Biotium Live-or-Dye™ 750/777 Fixable Viability Staining Kit (200 assays)
Supplier:  Biotium 32008
Encompass_Preferred
Live-or-Dye Fixable Viability Staining Kits are designed for discrimination between live and dead cells during flow cytometry.
Catalog No. NC1642818