Learn More
Enzo Life Sciences Fixation, (buffer) (100 ml)
SDP
Catalog No. 50658672
Encompass_Preferred
Formulation: Liquid. PBS, pH 7.2, containing 4% formaldehyde. Applications: Flow Cytometry. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C
Catalog No. 50-658-672 Supplier Enzo Life Sciences Supplier No. ADI950011100
Formulation: Liquid. PBS, pH 7.2, containing 4% formaldehyde. Applications: Flow Cytometry. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C