Learn More
Med Vet International Catheter Fixation Device Stayfix Medium, 5 to 12 Fr., 680ME, Each
Catalog No. 503043536
Encompass_Preferred
Special stayfix design secures catheters in place without the use of sutures, reducing the exposure to sharps and sharps-related accidents. Provides safe and secure fixation for up to 7 days
Catalog No. 50-304-3536 Supplier Med Vet International Supplier No. 312463EA
Special stayfix design secures catheters in place without the use of sutures, reducing the exposure to sharps and sharps-related accidents. Provides safe and secure fixation for up to 7 days