Learn More
Biotium Fixation Permeabilization Other Buffers Buffers Accessories and Background Reducers Fixation Permeabilization and Other Buffers Chemical Reagents 50/PK
Catalog No. 5000501584
Encompass_Preferred
PBS (phosphate-buffered saline) buffer powder packet for preparing 2 L of 1X PBS buffer Box includes 50 packets
Catalog No. 50-005-01584 Supplier Biotium Supplier No. 22033
PBS (phosphate-buffered saline) buffer powder packet for preparing 2 L of 1X PBS buffer Box includes 50 packets