Learn More
Biotium Fixation Permeabilization Other Buffers DNA Gel Stains and Accessories DNA Gel Stains Chemical Reagents 50/PK
Catalog No. 5000501583
Encompass_Preferred
TAE (Tris-acetate-EDTA) buffer powder packet for preparing 1 L of 1X TAE buffer for nucleic acid gel electrophoresis Box includes 50 packets
Catalog No. 50-005-01583 Supplier Biotium Supplier No. 22031
Add to Cart
Add to Cart
TAE (Tris-acetate-EDTA) buffer powder packet for preparing 1 L of 1X TAE buffer for nucleic acid gel electrophoresis Box includes 50 packets