Learn More
Med Vet International Fixation Pins End Positive Thread, J0371H, Large, Each
Catalog No. 5000582029
Encompass_Preferred
Large. Shaft 5/32" dia., Thread 3/16" dia., Thread Port 11/2"L, Total 6"L, Thread Pitch 0.9mm
Catalog No. 50-005-82029 Supplier Med Vet International Supplier No. JOR371H
Large. Shaft 5/32" dia., Thread 3/16" dia., Thread Port 11/2"L, Total 6"L, Thread Pitch 0.9mm