Learn More
Electron Microscopy Sciences Fixative/Decalcifier 1 LT
SDP
Supplier:  Electron Microscopy Sciences 6414806
Encompass_Preferred
The solution fixes and decalcifies in one step. It prevents loss of cellular structure and enhances hematoxylin staining.
Catalog No. 50-259-39