Learn More
Electron Microscopy Sciences Fixative/Decalcifier 1 LT
SDP
Catalog No. 5025938
Encompass_Preferred
The solution fixes and decalcifies in one step. It prevents loss of cellular structure and enhances hematoxylin staining.
Catalog No. 50-259-38 Supplier Electron Microscopy Sciences Supplier No. 6414801
The solution fixes and decalcifies in one step. It prevents loss of cellular structure and enhances hematoxylin staining.