Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fixative-Free Lysing Solution, High-Yield Lyse

Catalog No. HYL250
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HYL250 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HYL250 Supplier Invitrogen™ Supplier No. HYL250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fixative-Free Lysing Solution, High-Yield Lyse

High-Yield Lyse is a premixed, fixative-free erythrocyte lysing solution that can be used to eliminate red cells from whole blood for flow cytometric analysis with minimal loss of rare blood cell populations. Using this reagent, lysis of erythrocytes is performed immediately following staining of the blood samples with Invitrogen's fluorochrome-conjugated monoclonal antibodies.

Related Links:

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.

Shelf life: 24 months from date of manufacture
Sample Type Blood
Product Type Lysing Solution
Quantity 500 mL
Reagent Type Whole Blood Lysis Buffer
Shipping Condition Room Temperature
What reagents can I use to lyse red blood cells other than the FIX & PERM Cell Permeabilization Kit?

Cal-Lyse premixed lysis solution (Cat. Nos. GAS010 and GAS010S100) is specifically formulated to lyse erythrocytes following monoclonal antibody staining of whole blood or bone marrow. Treatment with this reagent leads to both the lysis of red blood cells and the fixation of white cells. Treatment does not affect fluorophore-labeled antibodies bound to leukocytes, and leaves morphological scatter characteristics of leukocytes intact.
-High-Yield Lyse (Cat. No. HYL250) is a premixed, fixative-free erythrocyte-lysis solution that can be used to help eliminate red cells from whole blood for flow cytometric analysis with minimal loss of rare blood cell populations. Using this reagent, lysis of erythrocytes is performed immediately following staining of the blood samples with fluorophore-conjugated monoclonal antibodies; there is no need for a wash step. Both are IVD reagents.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.