Learn More
Ethos Biosciences Acrylic Mounting Medium, 16oz, Ethos
Catalog No. 5000574281
Encompass_Preferred
Acrylic Mounting Media is a clear, fast-drying mounting solution designed for labs needing slide preservation. Formulated to provide strong adhesion and long-term slide preservation, it is ideal for coverslipping stained specimens and routine slide preparation. The medium offers excellent optical clarity and minimal shrinkage, supporting reliable microscopic evaluation. Compatible with a wide range of staining protocols, it delivers consistent performance in clinical, research, and educational laboratories. Supplied in chemically compatible containers, Acrylic Mounting Media is suitable for routine and high-throughput laboratory workflows.
Catalog No. 50-005-74281 Supplier Ethos Biosciences Supplier No. 334916
Acrylic Mounting Media is a clear, fast-drying mounting solution designed for labs needing slide preservation. Formulated to provide strong adhesion and long-term slide preservation, it is ideal for coverslipping stained specimens and routine slide preparation. The medium offers excellent optical clarity and minimal shrinkage, supporting reliable microscopic evaluation. Compatible with a wide range of staining protocols, it delivers consistent performance in clinical, research, and educational laboratories. Supplied in chemically compatible containers, Acrylic Mounting Media is suitable for routine and high-throughput laboratory workflows.