Learn More
Anatech Ltd Z-Fix Concentrate, 1 gallon
Catalog No. NC9351419
Encompass_Preferred
Diluted Z-Fix Concentrate (Z-Fix) is designed to provide the desirable features of formaldehyde fixation without the negative effects of excessive crosslinking. Formaldehyde fixation is a two-step process. First, formaldehyde forms addition products with macromolecules. Second, the formaldehyde adduct crosslinks the macromolecules, firming the tissue. While necessary for complete fixation, the crosslinks (methylene bridges) can block access to antigenic epitopes. With Z-Fix, both the zinc and formalin initially attach to tissue sites. Additional zinc ions then form large coordination complexes that prevent the formaldehyde from further crosslinking by simply getting in the way. The zinc constituent of Z-Fix gives cellular components enough rigidity without undesirable hardening.
Catalog No. NC9351419 Supplier Anatech Ltd Supplier No. 171
Diluted Z-Fix Concentrate (Z-Fix) is designed to provide the desirable features of formaldehyde fixation without the negative effects of excessive crosslinking. Formaldehyde fixation is a two-step process. First, formaldehyde forms addition products with macromolecules. Second, the formaldehyde adduct crosslinks the macromolecules, firming the tissue. While necessary for complete fixation, the crosslinks (methylene bridges) can block access to antigenic epitopes. With Z-Fix, both the zinc and formalin initially attach to tissue sites. Additional zinc ions then form large coordination complexes that prevent the formaldehyde from further crosslinking by simply getting in the way. The zinc constituent of Z-Fix gives cellular components enough rigidity without undesirable hardening.