Learn More
Fine Science Tools Magnetic Fixator Retraction System, Aluminum, 3 cm high
SDP
Catalog No. NC9162717
Encompass
Product: Magnetic Fixator Retraction System - Short Fixator / Item: Short Fixator/ Alloy/Material: Aluminum/ Dimensions: 2.5cm High
Catalog No. NC9162717 Supplier Fine Science Tools Supplier No. 1820001
Product: Magnetic Fixator Retraction System - Short Fixator / Item: Short Fixator/ Alloy/Material: Aluminum/ Dimensions: 2.5cm High