Learn More
Perkin Elmer US LLC Fixed 1.0 mm Injector UHP Quartz Torch (White/White/White)
Catalog No. 502112685
Encompass_Preferred
The one-piece quartz torch has the injector integrated directly into the torch.
Catalog No. 50-211-2685 Supplier Perkin Elmer US LLC Supplier No. N8152600
Add to Cart
Add to Cart
Request a Quote
The one-piece quartz torch has the injector integrated directly into the torch.