Learn More
Perkin Elmer US LLC Fixed 1.5 mm Injector UHP Quartz Torch (White/White)
Catalog No. 502111544
Encompass_Preferred
The one-piece quartz torch has the injector integrated directly into the torch.
Catalog No. 50-211-1544 Supplier Perkin Elmer US LLC Supplier No. N8152601
The one-piece quartz torch has the injector integrated directly into the torch.