Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Fixed Benchtop Organizer

Catalog No. 22505340
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-505-340 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-340 Supplier Marketlab Supplier No. 9952
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Storage organizer with angled dividers to help keep supplies organized, maximize space, and provide easy access.

  • Small footprint maximizes your bench space
  • Designed for easy supply stocking and retrieval
  • At-a-glance inventory assessment
  • Durable PVC construction yet lightweight and easy to move

Specifications

Product Type Organizer
Color White
Material PVC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.