Learn More
Active Motif Fixed Cell ATAC-Seq Kit 16 rxn
SDP
Catalog No. NC2586638
Encompass
ATAC-Seq is a rapid assay that enables analysis of epigenetic profiles across the genome by identification of regions that have open or accessible chromatin states. Active Motif's Fixed Cell ATAC-Seq Kit provides the reagents necessary to produce 16 unique sequencing-ready Illumina-compatible ATAC-Seq libraries from 50,000-100,000 formaldehyde-fixed cells per reaction. The optimized protocol guides you through the steps for sample preparation, tagmentation and library preparation, yielding Next-gen sequencing-ready libraries that can be multiplexed in a single flow cell sequencing run.
Catalog No. NC2586638 Supplier Active Motif Supplier No. 53151
Add to Cart
Edge
Add to Cart
ATAC-Seq is a rapid assay that enables analysis of epigenetic profiles across the genome by identification of regions that have open or accessible chromatin states. Active Motif's Fixed Cell ATAC-Seq Kit provides the reagents necessary to produce 16 unique sequencing-ready Illumina-compatible ATAC-Seq libraries from 50,000-100,000 formaldehyde-fixed cells per reaction. The optimized protocol guides you through the steps for sample preparation, tagmentation and library preparation, yielding Next-gen sequencing-ready libraries that can be multiplexed in a single flow cell sequencing run.