Learn More
Cole-Parmer Polypropylene dual-blade paddle assembly; 5/16" x 13-13/16"L, 1-5/8" paddle dia
Catalog No. NC9725589 Shop All Cole-Parmer Products
Encompass_Preferred
For mixing of medium viscosity solutions.
This item is not returnable. View return policy
Catalog No. NC9725589 Supplier Cole-Parmer Supplier No. 0636710
For mixing of medium viscosity solutions.