Learn More
Bellco Glass, Inc. Fixed Frame Rllr App 10DK,115V 50 Position
SDP
Supplier:  Bellco Glass, Inc. 763000510
Encompass_Preferred
The Bellco Cell Production Roller Apparatus has been designed for production of animal cells, viruses and cell products. Modular units are available from 5 to 45 bottle positions and allow easy expansion with modular add-on decks. Non-modular production units come in 45 and 50 bottle positions with over 80,000 cm2 of surface area. Rollers have a key design to eliminate slipping and are easily removable for servicing of bearings. Rollers are formulated from a chemical-resistant non-slip EDPM. All units feature a solid state control with heavy-duty DC motor and analog tachometer. Non-modular production models feature rubber bumpers, handles and a slotted rear guard. Optional All Position Drives are available for use with plastic roller bottles to prevent slipping of the light-weight bottles. An optional Stand By Drive and Alarm System provides motion and power failure alarms and has a redundant drive system that automatically drives the unit should the primary fail. MADE IN THE USA
Catalog No. 50-194-5900
Add to Cart
Request a Quote