Learn More
Electron Microscopy Sciences Fixed Handle 24 Slide Rack Only
SDP
Catalog No. 50931048
Encompass_Preferred
Fixed Handle Rack: A stainless steel rack with a fixed handle for use with large slides. The rack is grooved to take 76 mm microscope slides up to 50 mm wide horizontally. The rack accommodates up to 24. Vertical Slide Rack: A stainless steel rack with a fixed handle and grooves to take 76 x 26 mm microscope slides vertically for maximum use of space and solution. The rack accommodates up to 48 slides (24 slides each row).
Catalog No. 50-931-048 Supplier Electron Microscopy Sciences Supplier No. 71421FR
Fixed Handle Rack: A stainless steel rack with a fixed handle for use with large slides. The rack is grooved to take 76 mm microscope slides up to 50 mm wide horizontally. The rack accommodates up to 24. Vertical Slide Rack: A stainless steel rack with a fixed handle and grooves to take 76 x 26 mm microscope slides vertically for maximum use of space and solution. The rack accommodates up to 48 slides (24 slides each row).