Learn More
MSC Vikan Hand Squeegee, Polypropylene, 9.8 in Length, FDA Compliant
Catalog No. 502835514
Encompass
This hand squeegee effectively removes water from surfaces. It features a double-bladed design and comes with an easy-to-remove replacement cassette for convenient blade changes.
  • Effectively removes water from surfaces.
  • Features a double-bladed design.
  • Includes an easy-to-remove replacement cassette.
  • Constructed from polypropylene and cellular rubber.
  • Complies with FDA regulations.
  • Equipped with a 5.51 in handle.
Catalog No. 50-283-5514 Supplier MSC Supplier No. 47166962
This hand squeegee effectively removes water from surfaces. It features a double-bladed design and comes with an easy-to-remove replacement cassette for convenient blade changes.
  • Effectively removes water from surfaces.
  • Features a double-bladed design.
  • Includes an easy-to-remove replacement cassette.
  • Constructed from polypropylene and cellular rubber.
  • Complies with FDA regulations.
  • Equipped with a 5.51 in handle.