Learn More
MSC Vikan Floor Squeegee, Replaceable Cassette, 700 mm, White
Catalog No. 502839755
Encompass
This squeegee features a double-lipped cellular rubber cassette designed for comprehensive cleaning tasks. It is compatible with standard handles, and its cassette can be easily removed for cleaning and replacement.
  • Designed for all types of cleaning.
  • Cassette is easily removed for cleaning and replacement.
  • Ideal for food production, food stores, restaurants, and foodservice.
  • Flexible blade follows surface contours, improving water and dirt removal.
  • Double cellular rubber blades are flexible, waterproof, and highly effective.
  • Most effective for quickly removing large quantities of water.
  • Easy to clean and maintain, promoting good hygiene.
  • Durable construction for a long lifespan.
  • Resistant to chemicals and cleaning agents.
  • Compatible with Euro-thread handles for secure attachment.
Catalog No. 50-283-9755 Supplier MSC Supplier No. 78032976
This squeegee features a double-lipped cellular rubber cassette designed for comprehensive cleaning tasks. It is compatible with standard handles, and its cassette can be easily removed for cleaning and replacement.
  • Designed for all types of cleaning.
  • Cassette is easily removed for cleaning and replacement.
  • Ideal for food production, food stores, restaurants, and foodservice.
  • Flexible blade follows surface contours, improving water and dirt removal.
  • Double cellular rubber blades are flexible, waterproof, and highly effective.
  • Most effective for quickly removing large quantities of water.
  • Easy to clean and maintain, promoting good hygiene.
  • Durable construction for a long lifespan.
  • Resistant to chemicals and cleaning agents.
  • Compatible with Euro-thread handles for secure attachment.