Learn More
MSC Vikan Floor Squeegee, Double Foam, 19.7 in, Red, 1 ea
Catalog No. 502870426
Encompass
This Vikan floor squeegee head with replacement cassette is designed to effectively return floors to a dry state. It features a classic double blade cassette that is flexible and functional, making it ideal for various flooring types and efficient for removing large amounts of water or food debris quickly.
  • Cellular double foam blade is flexible
  • Ideal for large amounts of water and food debris
  • Extra replacement cassette included
  • Color-coded to reduce potential for cross-contamination
  • Compatible with any handle (sold separately)
  • Body made with polypropylene
  • Cellular double foam rubber blade is removable and easy to clean
  • HACCP compliant due to color-coding
  • Autoclave safe
  • Dishwasher safe
Catalog No. 50-287-0426 Supplier MSC Supplier No. 47167036
This Vikan floor squeegee head with replacement cassette is designed to effectively return floors to a dry state. It features a classic double blade cassette that is flexible and functional, making it ideal for various flooring types and efficient for removing large amounts of water or food debris quickly.
  • Cellular double foam blade is flexible
  • Ideal for large amounts of water and food debris
  • Extra replacement cassette included
  • Color-coded to reduce potential for cross-contamination
  • Compatible with any handle (sold separately)
  • Body made with polypropylene
  • Cellular double foam rubber blade is removable and easy to clean
  • HACCP compliant due to color-coding
  • Autoclave safe
  • Dishwasher safe