Learn More
Chemglass Life Sciences Fixed Height Sample Tube for 15L BioReactors, M10 Thread, Max Immersion Length: 400mm
SDP
Catalog No. 501294444
Encompass
Fixed Height Sample Tube for 15L BioReactors, M10 Thread, Max Immersion Length: 400mm. Fixed height sampling tubes for bioreactors. Tubing is 1/4 inch OD.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4444 Supplier Chemglass Life Sciences Supplier No. CLS138526
Fixed Height Sample Tube for 15L BioReactors, M10 Thread, Max Immersion Length: 400mm. Fixed height sampling tubes for bioreactors. Tubing is 1/4 inch OD.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.