Learn More
Fritsch Milling And Sizing Inc Fixed Jaw P-1 Cl Size 1 Stainless Steel
SDP
Catalog No. 501534368
Encompass
Fixed crushing jaw, stainless steel for PULVERISETTE 1 classic line, Size 1 for the comminution of medium-hard, brittle samples
General material specifications:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples
Catalog No. 50-153-4368 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0030.10
Fixed crushing jaw, stainless steel for PULVERISETTE 1 classic line, Size 1 for the comminution of medium-hard, brittle samples
General material specifications:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples