Learn More
Fritsch Milling And Sizing Inc Fixed Jaw P-1 Pl+Cl Size 2 Stainless Steel
SDP
Catalog No. 501534386
Encompass
Fixed crushing jaw, stainless steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of medium-hard, brittle samples
General material specification:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples
Catalog No. 50-153-4386 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3030.10
Fixed crushing jaw, stainless steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of medium-hard, brittle samples
General material specification:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples