Learn More
Fritsch Milling And Sizing Inc Fixed Jaw P-1 Pl+Cl Size 2 Hardmetal Tungsten Carbide
SDP
Catalog No. 501534388
Encompass
Fixed crushing jaw, hardmetal tungsten carbide for PULVERISETTE 1 premium line and classic line, Size 2
General material specifications:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: very good
Suitable for: hard, abrasive samples
Catalog No. 50-153-4388 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3050.08
Add to Cart
Add to Cart
Request a Quote
Fixed crushing jaw, hardmetal tungsten carbide for PULVERISETTE 1 premium line and classic line, Size 2
General material specifications:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: very good
Suitable for: hard, abrasive samples