Learn More
Trajan Scientific FIXED NEEDLE SYRINGE
SDP
Supplier:  Trajan Scientific 0029871
Encompass_Preferred
Trajan Thermo Scientific AS2000 Autosampler Syringe, Diameter Outer Needle: 0.63 mm, Length Needle: 50 mm, Model: 10F-BT-GT-5/0.63C, Needle Gauge: 23 G, Volume: 10 uL, Connection Type: Fixed, For Use With: Gas chromatography, Thermo Scientific instruments AS2000, Needle Point Style: Cone
Catalog No. 50-253-0354