Learn More
Trajan Scientific FIXED NEEDLE SYRINGE 6/PK
SDP
Catalog No. 502530363
Encompass_Preferred
Trajan Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments, Diameter Outer Needle: 0.47 mm, 0.018 in., Length Needle: 50 mm, 1.96 in., Model: 10F-BT-GT-5/0.47C, Needle Gauge: 26 G, Volume: 10 uL, Connection Type: Fixed
Catalog No. 50-253-0363 Supplier Trajan Scientific Supplier No. 0029761
Trajan Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments, Diameter Outer Needle: 0.47 mm, 0.018 in., Length Needle: 50 mm, 1.96 in., Model: 10F-BT-GT-5/0.47C, Needle Gauge: 26 G, Volume: 10 uL, Connection Type: Fixed