Learn More
Trajan Scientific FIXED NEEDLE SYRINGE
SDP
Supplier:  Trajan Scientific 0039871
Encompass_Preferred
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 25 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 50 mm, Connection Type: Fixed, For Use With: Gas chromatography, Antek instruments, Length: 50 mm, Model: 25F-BT-GT-0.63C
Catalog No. 50-253-0344