Learn More
Trajan Scientific FIXED NEEDLE SYRINGE
SDP
Supplier:  Trajan Scientific 002967
Encompass_Preferred
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.5 mm, Length Needle: 50 mm, Connection Type: Fixed, For Use With: Gas chromatography, CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus & AS3000/2000/200/800
Catalog No. 50-253-0367