Learn More
McKesson General Medical FIXED SPEED CENTRIFUGE 3400RPM
Catalog No. NC2190964
Encompass_Preferred
Fixed Speed Centrifuge McKesson 6 Place Fixed Angle Rotor Fixed speed at 3,400 RPM.McKesson Fixed Speed CentrifugeSix place rotor for use with standard 5 to 10 ml tubes. Can spin up to three 15 ml tubes; 2 ml and 4-5 ml tubes can be spun with available adapters.Fixed speed at 3,400 RPM.Dependable mechanical 30-minute timer with bell signal at end of run.High impact plastic housing for years of dependable use.Safety switch in lid shuts off motor when lid is unlatched. Latch release located on the right front side of unit.Power indicator light tells when unit is powered on.Not Made with Natural Rubber Latex.
This item is not returnable. View return policy
Catalog No. NC2190964 Supplier McKesson General Medical Supplier No. 1047221
Fixed Speed Centrifuge McKesson 6 Place Fixed Angle Rotor Fixed speed at 3,400 RPM.McKesson Fixed Speed CentrifugeSix place rotor for use with standard 5 to 10 ml tubes. Can spin up to three 15 ml tubes; 2 ml and 4-5 ml tubes can be spun with available adapters.Fixed speed at 3,400 RPM.Dependable mechanical 30-minute timer with bell signal at end of run.High impact plastic housing for years of dependable use.Safety switch in lid shuts off motor when lid is unlatched. Latch release located on the right front side of unit.Power indicator light tells when unit is powered on.Not Made with Natural Rubber Latex.