Learn More
Drummond™ Fixed-Volume Microdispensers
Catalog No. 2117010
Change view
Click to view available options
Volume (Metric):
10 μL
100 μL
100 to 200 μL
20 μL
25 μL
250 μL
5 μL
50 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-170-10 5 μL
21-176E 100 μL
21-176A 10 μL
21-176F 100 to 200 μL
21-176C 25 μL
21-176B 20 μL
21-176D 50 μL
21-172 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 21-170-10 Supplier Drummond™ Supplier No. 3000105
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Overall volumetric accuracy of ±1% at full volume
  • Contain borosilicate glass bores with stainless steel plungers, except the 1000μL unit which has precision-molded plastic bores
  • All plungers with capacities 25μL and larger have PTFE tips
  • The 5μL unit (Cat. No. 21-170-10) will deliver 1 to 5μL
Order Info
Repeat Kit is available for 1000μL unit. Custom spacers are available permitting fractional deliveries on all units; contact your Fisher Scientific™ Customer Service Representative.
Specifications
Accuracy ±1%
Number of Channels 1
Type Fixed Volume Microdispenser
Volume (Metric) 5 μL
For Use With (Application) Precision molded Plastic bores
Includes 2 Plungers, 2 O-rings, 1 collet, 1 coil spring, 1 spring guide