Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Fixed Volume Mini Pipettors
SCI_ED

Catalog No. S012413 Shop All Fischer Technical Company Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
Color:
Pink
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
S012413 10 μL White
S012412 20 μL Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S012413 Supplier Fischer Technical Company™ Supplier No. 347020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fixed Volume Mini Pipettors offer accuracy at an affordable price.

  • Unique miniature size pipettors provide accurate fixed volume dispensing
  • Ideal for micro-volumes and gel loading applications
  • Accept universal fit disposable pipette tips
  • Accepts 200 μL tips
  • Available in two sizes, color-coded for easy identification
  • Accuracy 0.3 μL and 0.6 μL, respectively

Specifications

Autoclavable Autoclavable
Color White
Disposable Reusable
For Use With Micropipette Tip
Material Polypropylene
Model D590
Number of Channels 1
Sterility Non-sterile
Type Fixed Volume Minipipette
Volume (Metric) 10 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.