Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Fixed Volume MiniPipets
SDPSCI_ED

Catalog No. S90244B
Click to view available options
Volume (Metric):
10 μL
100 μL
20 μL
25 μL
30 μL
50 μL
75 μL

Easy-to-use, precise operation and ergonomic design

Robust, accurate, easy to use, color coded, fun & cost effective micropipets which use standard micropipet tips. No need to calibrate and impossible to measure the wrong volume!

Warranty and Services

Lifetime

Specifications

Compatible Tips Standard micropipet tip
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Standard Micropipette Tip
Material Plastic
Model Fix 20 μL
Number of Channels 1
Sterility Non-sterile
Type Fixed Volume Minipipet
Volume (Metric) 20 μL
For Use With (Equipment) Standard micropipet tip
Warranty Lifetime
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.