Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Fixed Volume MiniPipets
SDPSCI_ED

Catalog No. p-3032138 Shop All EDVOTEK Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
30 μL
50 μL
75 μL
100 μL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S90244H 75 μL
S90244J 100 μL
S90244A 10 μL
S90244B 20 μL
S90244G 50 μL
S90244D 30 μL
S90244C 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S90244H Supplier Edvotek™ Supplier No. 5882
Add to Cart
Add to Cart

Easy-to-use, precise operation and ergonomic design

  • Utilize standard disposable micropipet tips
  • Lifetime warranty
  • Cost-effective

Warranty and Services

Lifetime

Specifications

Compatible Tips Standard micropipet tip
Autoclavable Not Autoclavable
Color Gray
Disposable Single-use
For Use With Standard Micropipette Tip
Material Plastic
Number of Channels 1
Sterility Non-sterile
Type Fixed Volume Minipipet
Volume (Metric) 75 μL
For Use With (Equipment) Standard micropipet tip
Warranty Lifetime
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.