Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Fixed Volume MiniPipettes™
SDPSCI_ED

Catalog No. S05658 Shop All EDVOTEK Products
Change view
Click to view available options
Volume (Metric):
35 μL
40 μL
Color:
Blue
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
S05658 35 μL Blue
S05659 40 μL Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S05658 Supplier Edvotek™ Supplier No. 5872
Add to Cart
Add to Cart
This item is not returnable. View return policy

Robust, color-coded micropipettes are cost-effective and use standard micropipette tips. No need to calibrate, making it nearly impossible to measure the wrong volume.

  • Fun, easy to use, and color-coded
  • Ideal for beginners or middle school students

Specifications

Compatible Tips Standard Micropipette Tips
Autoclavable Not Autoclavable
Color Blue
Disposable Reusable
For Use With Beginners
Material Plastic
Model 587-2
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Fixed Volume
Volume (Metric) 35 μL
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.