Learn More
Description
- Fun, easy to use, and color-coded
- Ideal for beginners or middle school students
Specifications
Specifications
| Compatible Tips | Standard Micropipette Tips |
| Autoclavable | Not Autoclavable |
| Color | Blue |
| Disposable | Reusable |
| For Use With | Beginners |
| Material | Plastic |
| Model | 587-2 |
| Number of Channels | 1 |
| Programmability | No |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

