Learn More
Hach Company Fixed Volume, Micropipette, 1000 µl
Supplier:  Hach Company 2946010
Encompass
Micropipette features a two-stage plunger dispenser, which ensures high accuracy and precision in pipetting techniques and facilitates last drop dispensing. Unique tip cone accepts both micro and ultra micro pipet tips. Each micropipette is fully autoclavable and color-coded for easy identification. Includes one pipet tip. Additional pipet tips sold separately.
Catalog No. 50-199-6914