Learn More
Cole-Parmer Cole-Parmer Fixed-Volume Lightweight Metal Pipette, Blue, 5000 uL; Each
Catalog No. NC1258314 Shop All Cole-Parmer Products
Encompass_Preferred
For accurate and consistent results, choose a fixed-volume pipettor. Ideal for repetitive delivery of reagents and solutions. These durable lightweight pipettors feature a precision-machined plunger and barrel assembly. The internal polyurethane multi-ring seals prevent fluid leakage far superior to single O-ring pipettors. Pipettors are color-coded to help you keep track of the proper dispensing volume — an excellent choice for a busy lab! Autoclavable
Catalog No. NC1258314 Supplier Cole-Parmer Supplier No. 0783976
For accurate and consistent results, choose a fixed-volume pipettor. Ideal for repetitive delivery of reagents and solutions. These durable lightweight pipettors feature a precision-machined plunger and barrel assembly. The internal polyurethane multi-ring seals prevent fluid leakage far superior to single O-ring pipettors. Pipettors are color-coded to help you keep track of the proper dispensing volume — an excellent choice for a busy lab! Autoclavable