Learn More
Waters Corp Fixed Wash Seal, .125 ID, 2/pk
Supplier:  Waters Corp 700011663
Encompass_Preferred
This replacement fixed wash seal features a .125 ID and is supplied in a 2-pack. It is specifically designed for use in ACQUITY Arc Bio Quaternary Solvent Managers, ensuring optimal performance and compatibility.
  • Designed for UPLC systems
  • Provided as a pack of two units
  • Intended for annual replacement
  • Essential component for ACQUITY Arc Bio Quaternary Solvent Managers
Catalog No. 50-239-2469