Learn More
Aviva Systems Biology Corp FJX1 Antibody - N-terminal region (ARP47013_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47013_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FJX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FJX1. Alternate names: FLJ22416, FLJ25593ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-59450