Learn More
TARGETMOL CHEMICALS INC FK-330 1MG
SDP
Catalog No. 502877852
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. FK-330 (FR-260330) is a novel orally active inducible nitric oxide synthase inhibitor with potential anticancer and antitumor activity that prevents ischemia and reperfusion injury in rat liver transplantation. Purity 99.89%
Catalog No. 50-287-7852 Supplier TARGETMOL CHEMICALS INC Supplier No. T317951MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. FK-330 (FR-260330) is a novel orally active inducible nitric oxide synthase inhibitor with potential anticancer and antitumor activity that prevents ischemia and reperfusion injury in rat liver transplantation. Purity 99.89%