Learn More
Selleck Chemical LLC FK-3311 S2595-5mg
SDP
Catalog No. 503112369
Encompass_Preferred
FK-3311 (COX-2 Inhibitor V) is a selective cell-permeable and orally available inhibitor of cyclooxygenase-2 (COX-2) with antiinflammatory FK-3311 has protective effects against hepatic warm ischemia-reperfusion injury by marked inhibition of TxA2
Catalog No. 50-311-2369 Supplier Selleck Chemical LLC Supplier No. S25955MG
FK-3311 (COX-2 Inhibitor V) is a selective cell-permeable and orally available inhibitor of cyclooxygenase-2 (COX-2) with antiinflammatory FK-3311 has protective effects against hepatic warm ischemia-reperfusion injury by marked inhibition of TxA2