Learn More
Medchemexpress LLC FK 3311 (COX-2 inhibitor V) | 116686-15-8 | MFCD00876455 | 98.7% | 341.33 g·mol⁻1 | C15H13F2NO4S | 5 MG
SDP
Catalog No. 5000221050
Encompass_Preferred
FK 3311 is a selective cyclooxygenase-2 (COX-2) inhibitor supplied as a powdered research reagent for studies of inflammation and COX-2-mediated pathways. Analytical characterization and stability data are provided to support assay and in vivo use.
  • Selective COX-2 inhibition for inflammation research.
  • High reported purity (≈98.7%) suitable for biochemical assays.
  • Characterized by NMR and LC-MS with available COA and datasheet.
  • Powder form with defined storage conditions to maintain stability.
  • Suitable for in vitro and in vivo pharmacology studies.
Catalog No. 50-002-21050 Supplier Medchemexpress LLC Supplier No. HY144455MG
FK 3311 is a selective cyclooxygenase-2 (COX-2) inhibitor supplied as a powdered research reagent for studies of inflammation and COX-2-mediated pathways. Analytical characterization and stability data are provided to support assay and in vivo use.
  • Selective COX-2 inhibition for inflammation research.
  • High reported purity (≈98.7%) suitable for biochemical assays.
  • Characterized by NMR and LC-MS with available COA and datasheet.
  • Powder form with defined storage conditions to maintain stability.
  • Suitable for in vitro and in vivo pharmacology studies.