Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FK 453 | 121524-18-3 | 99.2% | 375.46 | C23H25N3O2 | 25 MG
SDP

Catalog No. 5000244093
Encompass_Preferred

FK 453 is a research-use adenosine A1 receptor antagonist (compound 1d) characterized by potent activity and a defined chemical profile. It exhibits an IC50 of 17.2 nM and is supplied as a high-purity powder with recommended storage conditions for both powder and solution forms.

  • Potent, selective adenosine A1 receptor antagonist (IC50 = 17.2 nM).
  • High purity: 99.2%.
  • Molecular formula C23H25N3O2 and molecular weight 375.46.
  • Available in multiple small-scale package sizes, including 25 mg.
  • Storage recommended as powder at -20°C (3 years) or 4°C (2 years).
  • In solvent storage guidance: -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-44093 Supplier Medchemexpress LLC Supplier No. HY105275A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FK 453 is a research-use adenosine A1 receptor antagonist (compound 1d) characterized by potent activity and a defined chemical profile. It exhibits an IC50 of 17.2 nM and is supplied as a high-purity powder with recommended storage conditions for both powder and solution forms.

  • Potent, selective adenosine A1 receptor antagonist (IC50 = 17.2 nM).
  • High purity: 99.2%.
  • Molecular formula C23H25N3O2 and molecular weight 375.46.
  • Available in multiple small-scale package sizes, including 25 mg.
  • Storage recommended as powder at -20°C (3 years) or 4°C (2 years).
  • In solvent storage guidance: -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.