Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fk-453 | 121524-18-3 | 99.2% | 375.46 g·mol⁻¹ | C23H25N3O2 | 100 MG
SDP

Catalog No. 5000243686
Encompass_Preferred

FK 453 is a potent, selective adenosine A1 receptor antagonist (IC50 = 17.2 nM) supplied as a high-purity small-molecule powder for research use. It includes recommended storage and solvent handling guidance to support reproducible in vitro assays.

  • High purity 99.2%.
  • Potent adenosine A1 receptor antagonist, IC50 17.2 nM.
  • Molecular weight 375.46 g·mol⁻¹; formula C23H25N3O2.
  • Recommended solvent DMSO at 100 mg/mL; may require ultrasonication.
  • Storage: powder -20°C (up to 3 years), 4°C (up to 2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Suitable for in vitro pharmacology and receptor binding studies.

Catalog No. 50-002-43686 Supplier Medchemexpress LLC Supplier No. HY105275A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FK 453 is a potent, selective adenosine A1 receptor antagonist (IC50 = 17.2 nM) supplied as a high-purity small-molecule powder for research use. It includes recommended storage and solvent handling guidance to support reproducible in vitro assays.

  • High purity 99.2%.
  • Potent adenosine A1 receptor antagonist, IC50 17.2 nM.
  • Molecular weight 375.46 g·mol⁻¹; formula C23H25N3O2.
  • Recommended solvent DMSO at 100 mg/mL; may require ultrasonication.
  • Storage: powder -20°C (up to 3 years), 4°C (up to 2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Suitable for in vitro pharmacology and receptor binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.